Titanium standard solution, for ICP, in HNO₃, CRM, VHG
Proveedor: LGC Standards PROMOCHEM
VHG-PTINF-100 VHG-LTINF-100 VHG-PTINF-50 VHG-PTINF-500 VHG-TTINF-100 VHG-TTINF-500
PROOVHG-PTINF-100EA 50 EUR
PROOVHG-PTINF-100 PROOVHG-LTINF-100 PROOVHG-PTINF-50 PROOVHG-PTINF-500 PROOVHG-TTINF-100 PROOVHG-TTINF-500
Titanium standard solution, for ICP, in HNO₃, CRM, VHG
Normativas Estándares catiónicos Estándares de titanio
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certificaciones: Traceable to NIST, SRM.