Titanium standard solution, for ICP, in H₂O, tr. F⁻ , CRM, VHG
Proveedor: LGC Standards PROMOCHEM
VHG-PTIW-100 VHG-PTIW-500 VHG-TTIW-100 VHG-TTIW-500
PROOVHG-PTIW-100EA 53 EUR
PROOVHG-PTIW-100 PROOVHG-PTIW-500 PROOVHG-TTIW-100 PROOVHG-TTIW-500
Titanium standard solution, for ICP, in H₂O, tr. F⁻ , CRM, VHG
Normativas Estándares catiónicos Estándares de titanio
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Certificaciones: Traceable to NIST, SRM.