Endothelial Cell Proliferation Medium Supplement-Mix, FCS
Proveedor: PROVITRO
2150001
PRTV2150001EA 124 EUR
PRTV2150001
Endothelial Cell Proliferation Medium Supplement-Mix, FCS
Suplementos y aditivos de cultivo celular
The supplement kit for endothelial cell proliferation medium is suitable for culturing Provitro human endothelial cells after adding the supplement kit components to 500 ml basal medium The formulation is optimised for initial seeding of 4000 cells/cm² up to confluence (approx. 90%). Feeder-layer, matrix substrates or other substances are not necessary. Due to the possibility of reduced proliferative activity we recommend to use the antibiotic supplement for freshly isolated cells only. The supplement kit for endothelial cell priliferation medium is thoroughly tested after each production. All components are tested in a stringent biological assay. Each batch is checked for human endothelial cells proliferating characteristics. The cells cultured in endothelial cell growth medium are checked regarding their morphology, the adherence rate, the colony forming efficiency and the population doubling time.
The most important functions of IFNG are the direct inhibition of viral replication, immunostimulation and immunomodulation. Its importance is underscored by its contribution to defense against viral, bacterial and tumor threats. Abnormal expression of IFN gamma is associated with autoimmune and autoinflammatory diseases. In addition, IFNG promotes the activity of natural killer cells, further emphasizing its central role in the immune system. Recombinant mouse IFNG, comprising 133 amino acids with a predicted 15,5 kDa mass, exhibits glycosylation-related bands of about 19,6 and 15,4 kDa in SDS-PAGE under reducing conditions.
Storage: Samples remain stable for a duration of twelve months from the receipt date when stored within the temperature range of −20 to −80 °C. To ensure optimal storage conditions, maintain a sterile environment at these temperatures. For improved preservation, it is advisable to aliquot the protein. Avoid subjecting the samples to repeated freeze-thaw cycles to prevent degradation and maintain the integrity of the material.
Precaución: Store the supplements at −20 °C. Do not use the supplements after expiry date.