IPTG (Isopropil-β-D-tiogalactopiranósido) enzyme substrate dioxane-free

Proveedor: Thermo Fisher Scientific

R0392 R0393
R0392EA 87.75 EUR
R0392 R0393
IPTG (Isopropil-β-D-tiogalactopiranósido) enzyme substrate dioxane-free
IPTG (Isopropil-β-D-tiogalactopiranosida)

The IPTG (isopropyl-beta-D-thiogalactopyranoside) is a highly stable synthetic analog of lactose


It inactivates the lac repressor and induces synthesis of beta-galactosidase, an enzyme that promotes lactose utilization. IPTG is used to induce expression of cloned genes under control of the lac operon It is used in conjunction with X-Gal to determine the lac phenotype in blue or white colony screening.

Formula: C₉H₁₈O₅S
Pto. Ebullición: 402 °C (1013 hPa)
Pto. de fusión: 109,5…110.5 °C
Temperatura de almacenaje: Congelador
Número MDL: MFCD00063273
Núm. CAS: 367-93-1
EINECS: 206-703-0

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR