Tubing cutting pliers
Proveedor: Restek
20193
REST20193EA 79 EUR
REST20193
Tubing cutting pliers
Alicates
1/16" tubing cutting pliers.
  • Cuts quickly, reducing distortion
  • Perfect for quick cuts for non-critical applications or in "hard to get at" places