Luer lock valve adaptors
Proveedor: Restek
21283
REST21283EA 152 EUR
REST21283
Luer lock valve adaptors
Racores y conectores
These Luer lock valve adaptors are suitable for connecting a Luer lock fitting to a Rheodyne or Valco valve.