Deionisers, parker hydroGen mate
Proveedor: Restek
22000
REST22000EA 5250 EUR
REST22000
Deionisers, parker hydroGen mate
Purificación de agua Desionizadores
This Parker HydroGen mate deionising water system is an economical means of providing deionized water to hydrogen generators or for other uses. It removes organics, phosphates, chlorine and essentially all ionizable constituents from water supply.
  • Quick and easy installation with minimal maintenance
  • Easy-fill dispensing gun
  • Visual indication for cartridge changes