Vacuum/pressure station, oil-free
Proveedor: Restek
24003 24016
REST24003EA 0 EUR
REST24003 REST24016
Vacuum/pressure station, oil-free
Bombas Bombas de vació
Vacuum/pressure station allows switching from pressure to vacuum in a matter of seconds. It is quiet and has a oil-free unit which will not contaminate the extraction system or samples. Provides 20" Hg (68 kPa) vacuum or 18 psi (124 kPa) pressure.