Manual microliter syringes, Hamilton 700 series
Proveedor: Restek
24536 24539 24967 20174 20175
REST24536EA 182 EUR
REST24536 REST24539 REST24967 REST20174 REST20175
Manual microliter syringes, Hamilton 700 series
Jeringas Jeringas para cromatografía
Each syringe plunger and barrel assembly is manufactured as one working unit.
  • With cemented or removable needles
  • Components are not interchangeable or individually replaceable
Información para pedidos: Replacement cemented or removable needles available separately.