Screw thread vials, 8-425
Proveedor: Restek
24619 24620 24621 24622
REST24619EA 38 EUR
REST24619 REST24620 REST24621 REST24622
Screw thread vials, 8-425
Viales
Screw thread vials fits all 2,0 ml, 12×32 mm vial based autosamplers and provide most secure environment for your sample because evaporation is minimised.