Adapters for capillary columns
Proveedor: Restek
24970 20544
REST24970EA 116 EUR
REST24970 REST20544
Adapters for capillary columns
Suministros de cromatografía Piezas de repuesto para cromatografía
These adapters are made of high quality stainless steel.
  • Ideal for Thermo Scientific TRACE and Focus SSL
  • Siltek®-treated version available for additional inertness