MA VPH matrix spike mix with surrogate (14 components)
Proveedor: Restek
30454
REST30454EA 79.5 EUR
REST30454
MA VPH matrix spike mix with surrogate (14 components)
Normativas Estándares de la industria petrolera
The MA VPH matrix spike mix consists of 14 components they are (C5) n-pentane, (C9) n-nonane, benzene, 2,5-dibromotoluene (SS), ethylbenzene, 2-methylpentane, methyl tert-butyl ether (mtbe), naphthalene, toluene, 1,2,4-trimethylbenzene, 2,2,4-trimethylpentane (isooctane), m-xylene, o-xylene and p-xylene.
Certificaciones: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.