8021/502.2 surrogate mix #1
Proveedor: Restek
30463
REST30463EA 52.5 EUR
REST30463
8021/502.2 surrogate mix #1
Normativas Estándar par análisis medioambiental
The mix consists of 1-bromo-2-chloroethane, 1-chloro-3-fluorobenzene and fluorobenzene.
Certificaciones: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.