DHA aromatics standard (37 components)
Proveedor: Restek
30729
REST30729EA 907 EUR
REST30729
DHA aromatics standard (37 components)
Normativas Estándares de la industria petrolera
This standard is produced for refineries performing Detailed Hydrocarbon Analysis (DHA) of crude oil feedstocks and fuels.
This standard contain 37 components: Benzene (8,255%), N-Butylbenzene (2,077%), sec-Butylbenzene (3,09%), tert-Butylbenzene (2,058%), tert-1-Butyl-4-Ethylbenzene (2,076%), tert-1-Butyl-2-Methylbenzene (2,077%), tert-1-Butyl-3,4,5-Trimethylbenzene (3,355%), 1,2-Dimethyl-3-Ethylbenzene (2,152%), 1,2-Dimethyl-4-Ethylbenzene (0,915%), 1,3-Dimethyl-2-Ethylbenzene (0,799%), 1,3-Dimethyl-5-Ethylbenzene (0,407%), 1,4-Dimethyl-2-Ethylbenzene (2,076%), Ethylbenzene (8,248%), Hexylbenzene (5,155%), Isobutylbenzene (3,099%), Isopropylbenzene (2,085%), 2-Methylbutylbenzene (0,118%).
It helps with calibrating complex hydrocarbon analyses and provides the greatest number of gravimetrically determined values for quantitative calibration.
Certificaciones: Meets requirements such as ASTM Methods D6729, D6730, and D6733. Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.