Capillary columns, Rtx®-35 (fused silica)
Proveedor: Restek
40402 40411 10420 10423 10424 10435 10437 10438 10439 10440 10453 10454 10455 10470 10485
REST40402EA 592 EUR
REST40402 REST40411 REST10420 REST10423 REST10424 REST10435 REST10437 REST10438 REST10439 REST10440 REST10453 REST10454 REST10455 REST10470 REST10485
Capillary columns, Rtx®-35 (fused silica)
Columnas de cromatografía
General-purpose columns for organochlorine pesticides, PCB congeners (e.g. Aroclor mixes), herbicides, pharmaceuticals, sterols, rosin acids, phthalate esters.
Midpolarity phase; Crossbond® diphenyl dimethyl polysiloxane
Temperature range: 40 to 320 °C
Similar phases: HP-35, DB-35, ZB-35.
Equivalent to USP G42 phase.