Analytical HPLC columns, Ultra Biphenyl (USP L11)
Proveedor: Restek
9109552 9109352 910950210 910950212 9109311 9109312 9109312-700 9109313-700 9109315 9109315-700 910931E 9109331 9109332 9109332-700 9109333-700 9109335 9109335-700 910933E 9109351 9109352-700 9109353-700 9109355 9109355-700 910935E 9109361 9109362 9109362-700 9109365 9109365-700 910936E 9109511 9109512 9109512-700 9109513-700 9109515 9109515-700 910951E 9109521 9109522 9109522-700 9109523-700 9109525 9109525-700 910952E 9109531 9109532 9109532-700 9109533-700 9109535 9109535-700 910953E 9109551 9109552-700 9109553-700 9109555 9109555-700 910955E 9109561 9109562 9109562-700 9109563-700 9109565 9109565-700 910956E 9109571 9109572 9109572-700 9109573-700 9109575 9109575-700 910957E
REST9109552EA 591 EUR
REST9109552 REST9109352 REST910950210 REST910950212 REST9109311 REST9109312 REST9109312-700 REST9109313-700 REST9109315 REST9109315-700 REST910931E REST9109331 REST9109332 REST9109332-700 REST9109333-700 REST9109335 REST9109335-700 REST910933E REST9109351 REST9109352-700 REST9109353-700 REST9109355 REST9109355-700 REST910935E REST9109361 REST9109362 REST9109362-700 REST9109365 REST9109365-700 REST910936E REST9109511 REST9109512 REST9109512-700 REST9109513-700 REST9109515 REST9109515-700 REST910951E REST9109521 REST9109522 REST9109522-700 REST9109523-700 REST9109525 REST9109525-700 REST910952E REST9109531 REST9109532 REST9109532-700 REST9109533-700 REST9109535 REST9109535-700 REST910953E REST9109551 REST9109552-700 REST9109553-700 REST9109555 REST9109555-700 REST910955E REST9109561 REST9109562 REST9109562-700 REST9109563-700 REST9109565 REST9109565-700 REST910956E REST9109571 REST9109572 REST9109572-700 REST9109573-700 REST9109575 REST9109575-700 REST910957E
Analytical HPLC columns, Ultra Biphenyl (USP L11)
Columnas de cromatografía
These Ultra biphenyl columns are made of high-purity, type-B silica that minimises activity and creates high-density bonding. These columns are designed for selective and reliable HPLC applications.
  • Substantial increases in retention, especially for dipolar, unsaturated, or conjugated solutes
  • Enhanced orthogonal selectivity when using methanol mobile phases
  • Ideal for increasing sensitivity and selectivity in LC-MS analyses
These columns provide a greater degree of dispersion than conventional phenyls and a greater degree of polarisability than phenylMore Product Information