HPLC silica columns, Viva Silica (USP L3)
Proveedor: Restek
9510513-700 9510515-700 9510563-700 9510573-700 9510575-700 9510515 9510575 951050210 951050212 9510511 9510512 9510512-700 951051E 9510521 9510522 9510522-700 9510523-700 9510525 9510525-700 951052E 9510531 9510532 9510532-700 9510533-700 9510535 9510535-700 951053E 9510551 9510552 9510552-700 9510553-700 9510555 9510555-700 951055E 9510561 9510562 9510562-700 9510565 9510565-700 951056E 9510571 9510572 9510572-700 951057E
REST9510513-700EA 0 EUR
REST9510513-700 REST9510515-700 REST9510563-700 REST9510573-700 REST9510575-700 REST9510515 REST9510575 REST951050210 REST951050212 REST9510511 REST9510512 REST9510512-700 REST951051E REST9510521 REST9510522 REST9510522-700 REST9510523-700 REST9510525 REST9510525-700 REST951052E REST9510531 REST9510532 REST9510532-700 REST9510533-700 REST9510535 REST9510535-700 REST951053E REST9510551 REST9510552 REST9510552-700 REST9510553-700 REST9510555 REST9510555-700 REST951055E REST9510561 REST9510562 REST9510562-700 REST9510565 REST9510565-700 REST951056E REST9510571 REST9510572 REST9510572-700 REST951057E
HPLC silica columns, Viva Silica (USP L3)
Columnas de cromatografía
These Viva columns are based on a wide pore material designed for optimal large-molecule separations. Base-deactivated spherical silica is useful for normal-phase or HILIC separations. These columns have a narrow distribution around the mean pore size, permitting a larger portion of the silica surface to play a role in the separation.
  • Rugged, spherical particles with 300 Å pore size
  • High proportion of pore/surface area available to large molecules
  • Excellent for separating peptides or proteins
Order Now
ESPECIFICACIONES
Environmentally Preferable
Extremo tapado N/A N/A
Rango de pH 2,5 - 8 2,5 - 8
Temperature limit 80 °C 80 °C
Código USP L3