Filter discs, biplane, series 16, porosity 0, VitraPOR®
Proveedor: ROBU GLASFILTER
16100 16400 16500 16600 16700 16900 16800 16200 16110 16120 16250 16300 62000 61900 61300 62800 62700 62900 62600 62500 62400 63800 63500 63000 61800 61700 61600 61500 61400 62100 62300 62200
ROBU16100EA 63.4 EUR
ROBU16100 ROBU16400 ROBU16500 ROBU16600 ROBU16700 ROBU16900 ROBU16800 ROBU16200 ROBU16110 ROBU16120 ROBU16250 ROBU16300 ROBU62000 ROBU61900 ROBU61300 ROBU62800 ROBU62700 ROBU62900 ROBU62600 ROBU62500 ROBU62400 ROBU63800 ROBU63500 ROBU63000 ROBU61800 ROBU61700 ROBU61600 ROBU61500 ROBU61400 ROBU62100 ROBU62300 ROBU62200
Filter discs, biplane, series 16, porosity 0, VitraPOR®
Filtros Discos de filtros
The unique porous structure of VitraPOR® sintered glass filters makes it flexible and versatile for many applications.
  • Very good chemical resistance
  • Very good temperature resistance
Made of borosilicate glass. The faces of filter discs from the series 16 are precisely ground down to tight tolerances and plane surfaces as may be required in specific installation circumstances.