Gas washing bottles, Drechsel pattern, VitraPOR®
Proveedor: ROBU GLASFILTER
40252 40254 40101 40100 40102 40103 40104 40250 40251 40253
ROBU40252EA 97.9 EUR
ROBU40252 ROBU40254 ROBU40101 ROBU40100 ROBU40102 ROBU40103 ROBU40104 ROBU40250 ROBU40251 ROBU40253
Gas washing bottles, Drechsel pattern, VitraPOR®
Frascos Botellas de lavado de gases
Borosilicate glass.
  • With sintered glass disc, porosity grade 0/1/2/3/4
  • All system components can be ordered separately
  • Excellent chemical and temperature resistance
Consists of the gas washing bottle with NS 29/32 standard ground joint socket and the insert with NS 29/32 cone joint and two glass tubing connectors with 11 mm external diameter.