TrueBlot® Anti-Goat IgG Magnetic Beads
Proveedor: Rockland Immunochemicals
00-1844-20 00-1844-50
ROCK00-1844-20EA 490 EUR
ROCK00-1844-20 ROCK00-1844-50
TrueBlot® Anti-Goat IgG Magnetic Beads
Perlas Bolas magnéticas
TrueBlot® Anti-goat IgG magnetic beads.
  • Used for separation and purification
  • immunoassays, immunoprecipitation, and IP western blots
  • Bead mean diameter is ~0,5 μm, bead concentration is 5 mg/ml
Trueblot® Magnetic beads are uniform, non-aggregating, super-paramagnetic beads consisting of a ferric oxide core functionalized with various silane groups. The super-paramagnetic nanoparticles are coupled with a biomolecule, such as rabbit anti-goat IgG, and are specifically designed, tested and quality controlled for isolation and purification of goat IgG,More Product Information
Order Now
ESPECIFICACIONES
Environmentally Preferable
Condiciones de envío Wet ice