Anti-DDB2 Rabbit Polyclonal Antibody

Proveedor: Rockland Immunochemicals
100-401-A10
ROCK100-401-A10EA 821 EUR
ROCK100-401-A10
Anti-DDB2 Rabbit Polyclonal Antibody
Anticuerpos
This antibody reacts with human DDB2 by western blot and immunoprecipitation.  The antibody immunoprecipitates in vitro translated Protein and Protein from cell lysates (using 293T, and others).  Coimmunoprecipitation of related Proteins has not been tested.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000;

Type: Primary
Antigen: DDB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno DDB2
Nombre del antígeno damage-specific DNA binding protein 2, 48kDa
Environmentally Preferable
Sinónimos de antígenos UV-DDB2|DDBB
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Unconjugated
Reactividad Human
Huésped Rabbit
ID del gen 1643
Isotipo
Western blot Yes
ELISA Yes
Precipitación inmunológica Yes
Epítopo C-terminal
Inmunógeno This antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 419-427 of Human DDB2 (C-terminal) coupled to KLH.
Número de aminoácidos 419 to 427
Formato Antiserum
Forma Liquid (sterile filtered)
Purification delipidation and defibrination
Concentración 85 mg/ml by Refractometry
Adsorción cruzada none
Storage buffer 0,01% (w/v) Sodium Azide
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR