Anti-ZBTB7B Rabbit Polyclonal Antibody

Proveedor: Rockland Immunochemicals
100-401-A21
ROCK100-401-A21EA 821 EUR
ROCK100-401-A21
Anti-ZBTB7B Rabbit Polyclonal Antibody
Anticuerpos
This antibody has been tested for use in ELISA, immunochemistry and western blotting. Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 80 kDa in size corresponding to cKrox by western blotting.

Recommended Dilutions: ELISA: 1:2500 - 1:10000; Western Blot: 1:500 - 1:3000; Immunochemistry: 1.25-2.5 µg/ml;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: ZBTB7B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Mouse, Human, Rat, Mouse
Order Now

ESPECIFICACIONES

Simbolo antígeno ZBTB7B
Nombre del antígeno zinc finger and BTB domain containing 7B
Environmentally Preferable
Sinónimos de antígenos Thpok|c-Krox|Zfp67
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Unconjugated
Reactividad Human,Rat,Mouse
Huésped Rabbit
ID del gen 22724
Isotipo
Western blot Yes
ELISA Yes
InmunoQuímica Yes
Epítopo Internal
Inmunógeno This antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal sequence of mouse cKrox Protein.
Formato Antiserum
Forma Lyophilized
Purification delipidated and defibrinated antiserum
Adsorción cruzada No
Storage buffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
Temperatura de almacenamiento –20 °C
Shipping temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR