Anti-CANX Rabbit Polyclonal Antibody

Proveedor: Rockland Immunochemicals
100-401-E97
ROCK100-401-E97EA 861 EUR
ROCK100-401-E97
Anti-CANX Rabbit Polyclonal Antibody
Anticuerpos
Anti-Calnexin -CT Antibody is suitable for use in WB. Expect a band approximately ~90kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CANX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR