Anti-PDCD4 Mouse Monoclonal Antibody (Unconjugated) [clone: 9G6]

Proveedor: Rockland Immunochemicals
200-301-B17
ROCK200-301-B17EA 4310 EUR
ROCK200-301-B17
Anti-PDCD4 Mouse Monoclonal Antibody (Unconjugated) [clone: 9G6]
Anticuerpos
This monoclonal antibody is suitable for ELISA, immunochemistry and western blotting.  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 62 kDa in size corresponding to phosphorylated Pdcd4 Protein.

Recommended Dilutions: ELISA: 1:400,000; Western Blot: 1:1000-1:3000; Immunochemistry: 1.25-2.5 µg/ml

Type: Primary
Antigen: PDCD4
Clonality: Monoclonal
Clone: 9G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat, Xenopod
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR