Anti-ABCA4 Mouse Monoclonal Antibody (Unconjugated) [clone: 3F4]

Proveedor: Rockland Immunochemicals
200-301-D05
ROCK200-301-D05EA 980 EUR
ROCK200-301-D05
Anti-ABCA4 Mouse Monoclonal Antibody (Unconjugated) [clone: 3F4]
Anticuerpos
Anti-ABCA4 antibody is suitable for use in ELISA, Western Blotting and IHC. Specific conditions should be optimized by the end user. Expect a band of ~250kDa in size corresponding to ABCA4 Proteins in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000; Immunochemistry: 1:100

Type: Primary
Antigen: ABCA4
Clonality: Monoclonal
Clone: 3F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Xenopus, Bovine
Order Now

ESPECIFICACIONES

Simbolo antígeno ABCA4
Environmentally Preferable
Nombre del antígeno ATP-binding cassette, sub-family A (ABC1), member 4
Sinónimos de antígenos Abcr|D430003I15Rik|Abc10|AW050280|RmP
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado Unconjugated
Clon 3F4
Reactividad Xenopus,Human,Bovine,Mouse
Huésped Mouse
ID del gen 11304
Isotipo IgG
Western blot Yes
ELISA Yes
InmunoQuímica Yes
Inmunógeno Anti-ABCA4 antibody was produced by repeated immunizations with partially purified bovine 220-kDa disc rim Protein.
Formato Antibody fraction
Forma Liquid
Molecular weight 220 kDa
Purification Protein G purified
Adsorción cruzada No
Storage buffer 0,01 M HEPES, 0,15 M Sodium Chloride, pH 7,50,1 mg/ml Bovine Serum Albumin - IgG and Protease free, 50% (v/v) Glycerol
Temperatura de almacenamiento –20 °C
Shipping temperature Dry Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR