Anti-GABRB3 Mouse Monoclonal Antibody (Unconjugated) [clone: S87-25]

Proveedor: Rockland Immunochemicals
200-301-F32
ROCK200-301-F32EA 841 EUR
ROCK200-301-F32
Anti-GABRB3 Mouse Monoclonal Antibody (Unconjugated) [clone: S87-25]
Anticuerpos
Anti-GABA-A receptor Beta3 Antibody is suitable for use in WB and IF microscopy. Expect a band approximately ~55kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Immunofluorescence: 1.0-10µg/ml; Western Blot: 1µg/ml; Immunochemistry: 0.1-1.0µg/ml

Type: Primary
Antigen: GABRB3
Clonality: Monoclonal
Clone: S87-25
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Human, Rat, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR