Anti-AKT1 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 5E5.F5.D7]

Proveedor: Rockland Immunochemicals
200-308-J34
ROCK200-308-J34EA 856 EUR
ROCK200-308-J34
Anti-AKT1 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 5E5.F5.D7]
Anticuerpos
Anti-AKT1 PE Antibody is suitable for Flow Cytometry, ELISA, immunochemistry, and western blotting. Expect a band approximately 56 kDa in size corresponding to AKT1 Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: Flow Cytometry: 1.0 µg/ml;
Restore with deionized water (or equivalent); 100 µl to 20 µl
Do not freeze! Light sensitive!

Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: 5E5.F5.D7
Conjugation: RPE (R-Phycoerythrin)
Epitope: Center
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse, Rat, Human
Order Now

ESPECIFICACIONES

Simbolo antígeno AKT1
Nombre del antígeno v-akt murine thymoma viral oncogene homolog 1
Environmentally Preferable
Sinónimos de antígenos CWS6|RAC|RAC-ALPHA|PRKBA|PKB|AKT|PKB-ALPHA
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado RPE (R-Phycoerythrin)
Clon 5E5.F5.D7
Reactividad Human,Rat,Mouse
Huésped Mouse
ID del gen 207
Isotipo IgG2a kappa
Western blot Yes
ELISA Yes
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Citometría de flujo Yes
Epítopo Internal
Inmunógeno Anti-AKT1 Antibody was produced in mice by repeated immunizations with a synthetic peptide corresponding to internal residues of human AKT1 Protein followed by monoclonal development.
Formato Antibody fraction
Forma Lyophilized
Purification Protein A purified
Concentración 1,0 mg/ml by UV absorbance at 280 nm
Adsorción cruzada none
Storage buffer 0,02 M Potassium Phosphate, 0,5 M Sodium Chloride, pH 7,2, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
Temperatura de almacenamiento Cool
Shipping temperature Ice
Tested applications Fluoresence ELISA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR