Anti-AKT3 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 9A5.H9.G7]

Proveedor: Rockland Immunochemicals
200-308-J36
ROCK200-308-J36EA 856 EUR
ROCK200-308-J36
Anti-AKT3 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 9A5.H9.G7]
Anticuerpos
Anti-AKT3 PE Antibody is suitable for Flow Cytometry, ELISA, immunochemistry, and western blotting. Expect a band approximately 56 kDa in size corresponding to AKT3 Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions:
Restore with deionized water (or equivalent); 100 µl to 20 µl
Do not freeze! Light sensitive!

Type: Primary
Antigen: AKT3
Clonality: Monoclonal
Clone: 9A5.H9.G7
Conjugation: RPE (R-Phycoerythrin)
Epitope: Center
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Mouse, Rat, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR