Anti-KTI3 Rabbit Polyclonal Antibody

Proveedor: Rockland Immunochemicals
200-401-079S
ROCK200-401-079SEA 226 EUR
ROCK200-401-079S
Anti-KTI3 Rabbit Polyclonal Antibody
Anticuerpos
This product has been assayed against 1.0 µg of Trypsin Inhibitor (Soybean) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat) and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: KTI3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Soy Bean
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR