Anti-RFX5 Rabbit Polyclonal Antibody

Proveedor: Rockland Immunochemicals
200-401-194
ROCK200-401-194EA 821 EUR
ROCK200-401-194
Anti-RFX5 Rabbit Polyclonal Antibody
Anticuerpos
Primary Rabbit Anti-RFX5 (aa 320 to 494) Reacts with Human, Mouse, Rat

Recommended Dilutions: ELISA: 1:5000 - 1:25000; GelShift: 0.5 µl to 1.0 µl; Western Blot: 1:1000;

Type: Primary
Antigen: RFX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR