Anti-CD80 Armenian Hamster Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 16-10A1]

Proveedor: Rockland Immunochemicals
200-B02-N89
ROCK200-B02-N89EA 644 EUR
ROCK200-B02-N89
Anti-CD80 Armenian Hamster Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 16-10A1]
Anticuerpos
Anti-CD80 is useful for immunochemistry, Immunofluorescence, Immunoprecipitation, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated).

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg); Sufficient to run approximately 500 tests
Do not freeze!

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: 16-10A1
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR