Anti-IgG (H+L) Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Proveedor: Rockland Immunochemicals
207-4202
ROCK207-4202EA 617 EUR
ROCK207-4202
Anti-IgG (H+L) Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticuerpos
Type: Secondary
Antigen: IgG (Golden Syrian Hamster) (Golden Syrian Hamster IgG)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)Epitope:
Host: Rabbit
Isotype: none
Reactivity: Hamster
Order Now

ESPECIFICACIONES

Simbolo antígeno IgG (H+L)
Environmentally Preferable
Tipo de anticuerpo Secundario
Clonalidad Polyclonal
Conjugado FITC (Fluorescein Isothiocyanate)
Reactividad Golden syrian hamster
Huésped Rabbit
Isotipo
Western blot Yes
ELISA Yes
InmunoQuímica Yes
Precipitación inmunológica Yes
FluorescenciaInmunológica Yes
Cambio de gel Yes
Citometría de flujo Yes
ChIP Yes
Neutralización Yes
Adsorción cruzada none
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR