Anti-IgM µ Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Proveedor: Rockland Immunochemicals
208-4307
ROCK208-4307EA 922 EUR
ROCK208-4307
Anti-IgM µ Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
This product has been assayed against 1.0 µg of Horse IgM in a standard capture ELISA using ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid)) as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 2.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: µ-chain
Host: Rabbit
Isotype: IgM
Reactivity: Horse
Order Now

ESPECIFICACIONES

Simbolo antígeno IgM µ
Environmentally Preferable
Tipo de anticuerpo Secundario
Clonalidad Polyclonal
Conjugado HRP (Horseradish Peroxidase)
Reactividad Horse
Huésped Rabbit
Isotipo
Western blot Yes
ELISA Yes
InmunoQuímica Yes
Precipitación inmunológica Yes
Cambio de gel Yes
Citometría de flujo Yes
ChIP Yes
Neutralización Yes
Inmunógeno Horse IgM mu heavy chain
Formato Antibody fraction
Forma Lyophilized
Purification multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above
Concentración 10,0 mg/ml by UV absorbance at 280 nm
Adsorción cruzada none
Storage buffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Gentamicin Sulfate. 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR