Anti-EBI3 Rat Monoclonal Antibody (Biotin) [clone: 10J811]

Proveedor: Rockland Immunochemicals
210-506-B66
ROCK210-506-B66EA 856 EUR
ROCK210-506-B66
Anti-EBI3 Rat Monoclonal Antibody (Biotin) [clone: 10J811]
Anticuerpos
This purified antibody has been tested for use in ELISA, Western Blot, and Flow Cytometry. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Flow Cytometry: 1.5-3.0µg/10x6 cells; Western Blot: 1µg/ml
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: EBI3
Clonality: Monoclonal
Clone: 10J811
Conjugation: Biotin
Epitope: Full length
Host: Rat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR