Anti-FITC Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Proveedor: Rockland Immunochemicals
600-103-096
ROCK600-103-096EA 1120 EUR
ROCK600-103-096
Anti-FITC Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Anti-Fluorescein Antibody Peroxidase Conjugated Suitable for immunoblotting (western or dot blot), ELISA, and in situ hybridization, immunoperoxidase electron microscopy and immunochemistry as well as other peroxidase-antibody based enzymatic assays

Recommended Dilutions: ELISA: 1:200,000; Western Blot: 1:5000 - 1:10000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Tag
Antigen: FITC
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR