Anti-HIS2AV Rabbit Polyclonal Antibody

Proveedor: Rockland Immunochemicals
600-401-914
ROCK600-401-914EA 821 EUR
ROCK600-401-914
Anti-HIS2AV Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:400 - 1:1600; Immunochemistry: 2 µg/ml;

Type: Primary
Antigen: HIS2AV
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Drosophila Melanogaster, Drosophila Melanogaster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR