Anti-SMAD3 Rabbit Polyclonal Antibody

Proveedor: Rockland Immunochemicals
600-401-920
ROCK600-401-920EA 821 EUR
ROCK600-401-920
Anti-SMAD3 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: ELISA: 1:10000 - 1:40000; Western Blot: 1:500 - 1:3000; Immunochemistry: 1:500 - 1:3000;

Type: Primary
Antigen: SMAD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Xenopus laevis, Xenopus Tropicalis, Zebrafish, Rat, Mouse, Pig, Bovine, Chicken
Order Now

ESPECIFICACIONES

Simbolo antígeno SMAD family member 3
Environmentally Preferable
Sinónimos de antígenos JV15-2|LDS3|HsT17436|HSPC193|LDS1C|MADH3
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Unconjugated
Reactividad Zebrafish,Pig,Xenopus,Xenopus tropicalis,Chicken,Human,Bovine,Rat,Mouse
Huésped Rabbit
ID del gen 4088
Isotipo IgG
Western blot Yes
ELISA Yes
InmunoQuímica Yes
Epítopo C-terminal
Inmunógeno This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to the C-terminal domain of human SMAD3 Protein.
Formato Antibody fraction
Forma Liquid (sterile filtered)
Purification affinity-purified
Adsorción cruzada No
Storage buffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
Temperatura de almacenamiento –20 °C
Shipping temperature Dry Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR