Anti-UBE2J1 Rabbit Polyclonal Antibody

Proveedor: Rockland Immunochemicals
600-401-991S
ROCK600-401-991SEA 226 EUR
ROCK600-401-991S
Anti-UBE2J1 Rabbit Polyclonal Antibody
Anticuerpos
This affinity purified antibody has been tested for use in ELISA, western blotting and immunoprecipitation.  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 35-40 kDa in size corresponding to Ube2j1 protein.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:200 - 1:2000; Immunoprecipitation: 1-2 µg;

Type: Primary
Antigen: UBE2J1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno UBE2J1
Nombre del antígeno ubiquitin-conjugating enzyme E2, J1
Environmentally Preferable
Sinónimos de antígenos HSPC205|UBC6|HSU93243|HSPC153|Ubc6p|NCUBE1|NCUBE-1|CGI-76|UBC6E
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Unconjugated
Reactividad Human
Huésped Rabbit
ID del gen 51465
Isotipo IgG
Western blot Yes
ELISA Yes
Precipitación inmunológica Yes
Inmunógeno This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to residues near the amino terminus of the human Ube2j1 Protein.
Formato Antibody fraction
Forma Liquid (sterile filtered)
Purification affinity-purified
Adsorción cruzada No
Storage buffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
Temperatura de almacenamiento –20 °C
Shipping temperature Dry Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR