Anti-TRIM29 Rabbit Polyclonal Antibody

Proveedor: Rockland Immunochemicals
600-401-B15
ROCK600-401-B15EA 821 EUR
ROCK600-401-B15
Anti-TRIM29 Rabbit Polyclonal Antibody
Anticuerpos
This affinity purified antibody has been tested for use in ELISA and western blotting.   Specific conditions for reactivity and detection of ATDC should be optimized by the end user.  Expect a band approximately ~66 kDa in size corresponding to ATDC.

Recommended Dilutions: ELISA: 1:100000; Western Blot: 1:500 to 3000;

Type: Primary
Antigen: TRIM29
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Human, Horse, Bovine, Chimpanzee, Macaque
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR