Anti-RFP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Proveedor: Rockland Immunochemicals
600-403-379
ROCK600-403-379EA 781 EUR
ROCK600-403-379
Anti-RFP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Polyclonal anti-RFP is designed to detect RFP and its variants. This antibody can be used to detect RFP by ELISA (sandwich or capture) for the direct binding of antigen.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:2000 - 1:5000; Immunochemistry: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 100 µl
Do not add Sodium Azide!

Type: Tag
Antigen: RFP
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR