Anti-HA Rabbit Polyclonal Antibody (DyLight® 649)

Proveedor: Rockland Immunochemicals
600-443-384
ROCK600-443-384EA 774 EUR
ROCK600-443-384
Anti-HA Rabbit Polyclonal Antibody (DyLight® 649)
Anticuerpos
Type: Tag
Antigen: HA
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 649
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan
Order Now

ESPECIFICACIONES

Simbolo antígeno HA
Environmentally Preferable
Tipo de anticuerpo Tag
Clonalidad Polyclonal
Conjugado DyLight® 649
Reactividad Mouse
Huésped Rabbit
Isotipo
Western blot Yes
ELISA Yes
InmunoQuímica Yes
FluorescenciaInmunológica Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR