Anti-IgG (H+L) Goat Polyclonal Antibody (Biotin)

Proveedor: Rockland Immunochemicals
603-106-002
ROCK603-106-002EA 272 EUR
ROCK603-106-002
Anti-IgG (H+L) Goat Polyclonal Antibody (Biotin)
Anticuerpos
Anti-Chicken IgG antibody is suitable for ELISA,Western Blot and Immunohistrochemistry applications. Specific conditions for reactivity should be optimized by end user.

Recommended Dilutions: ELISA: 1:800,000; Western Blot: 1:3000 - 1:30,000; Immunochemistry: 1:500 - 1:3000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR