Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 488)

Proveedor: Rockland Immunochemicals
609-141-123
ROCK609-141-123EA 231 EUR
ROCK609-141-123
Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 488)
Anticuerpos
The emission spectra for this DyLight® conjugate match the principle output wavelengths of most common fluorescence instrumentation.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR