Anti-IgG (H+L) Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Proveedor: Rockland Immunochemicals
609-4317
ROCK609-4317EA 400 EUR
ROCK609-4317
Anti-IgG (H+L) Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
Anti-Human IgG HRP is suitable for immunoblotting, western blot, dot blot, ELISA, immunoperoxidase electron microscopy and immunochemistry as well as other peroxidase-antibody based enzymatic assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:10000 - 1:150000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno IgG (H+L)
Environmentally Preferable
Tipo de anticuerpo Secundario
Clonalidad Polyclonal
Conjugado HRP (Horseradish Peroxidase)
Reactividad Human
Huésped Rabbit
Isotipo
Western blot Yes
ELISA Yes
InmunoQuímica Yes
Precipitación inmunológica Yes
Cambio de gel Yes
Citometría de flujo Yes
ChIP Yes
Neutralización Yes
Inmunógeno Anti-Human IgG (h&L) was produced by repeated immunization with human IgG whole molecule in rabbit.
Formato Antibody fraction
Forma Lyophilized
Purification purified by immunoaffinity chromatography
Concentración 2,0 mg/ml by UV absorbance at 280 nm
Adsorción cruzada Mouse
Storage buffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Gentamicin Sulfate. 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR