Anti-IgG (H+L) Goat Polyclonal Antibody (Biotin)

Proveedor: Rockland Immunochemicals
610-1619
ROCK610-1619EA 358 EUR
ROCK610-1619
Anti-IgG (H+L) Goat Polyclonal Antibody (Biotin)
Anticuerpos
Secondary Goat Anti-IgG (H&L) Reacts with Mouse

Recommended Dilutions: ELISA: 1:550000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR