Anti-IgG2b Rabbit Polyclonal Antibody (Cy3®)

Proveedor: Rockland Immunochemicals
610-404-042
ROCK610-404-042EA 468 EUR
ROCK610-404-042
Anti-IgG2b Rabbit Polyclonal Antibody (Cy3®)
Anticuerpos
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 500 µl

Type: Secondary
Antigen: IgG2b γ
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR