Anti-IgG (H+L) Rabbit Polyclonal Antibody (Texas Red®)

Proveedor: Rockland Immunochemicals
612-4926
ROCK612-4926EA 384 EUR
ROCK612-4926
Anti-IgG (H+L) Rabbit Polyclonal Antibody (Texas Red®)
Anticuerpos
Secondary Rabbit Anti-IgG (H&L) Reacts with Rat

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

ESPECIFICACIONES

Simbolo antígeno IgG (H+L)
Environmentally Preferable
Tipo de anticuerpo Secundario
Clonalidad Polyclonal
Conjugado Texas Red®
Reactividad Rat
Huésped Rabbit
Isotipo
Western blot Yes
ELISA Yes
InmunoQuímica Yes
Precipitación inmunológica Yes
FluorescenciaInmunológica Yes
Cambio de gel Yes
Citometría de flujo Yes
ChIP Yes
Neutralización Yes
Inmunógeno Rat IgG whole molecule
Formato Antibody fraction
Forma Lyophilized
Purification purified by immunoaffinity chromatography
Concentración 1,0 mg/ml by UV absorbance at 280 nm
Adsorción cruzada Human
Storage buffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR