Anti-IgG (H+L) Donkey Polyclonal Antibody (AP (Alkaline Phosphatase))

Proveedor: Rockland Immunochemicals
612-705-002
ROCK612-705-002EA 341 EUR
ROCK612-705-002
Anti-IgG (H+L) Donkey Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticuerpos
This product has been assayed against 1.0 µg of Rat IgG in a standard capture ELISA using pNPP p-nitrophenyl phosphate code # NPP-10 as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Heavy and Light chain
Host: Donkey
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR