Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (Texas Red®)

Proveedor: Rockland Immunochemicals
613-4904
ROCK613-4904EA 384 EUR
ROCK613-4904
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (Texas Red®)
Anticuerpos
Secondary Rabbit Anti-F(ab')2 Reacts with Sheep (Ovine)

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR