Anti-IgG (H+L) Goat Polyclonal Antibody (Biotin)

Proveedor: Rockland Immunochemicals
706-106-002
ROCK706-106-002EA 431 EUR
ROCK706-106-002
Anti-IgG (H+L) Goat Polyclonal Antibody (Biotin)
Anticuerpos
This product has been assayed against 1.0 µg of Guinea Pig IgG in a standard capture ELISA using Peroxidase Conjugated Streptavidin and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid)) as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR